Learn More
Biotium 5-(AND-6)-CARBOXYRhodamine 110-X, SUCCIN
Supplier:  Biotium 90003
Encompass_Preferred
5-(and-6)-Carboxyrhodamine 110-X, succinimidyl ester (5(6)-CR 110-X, SE) readily couples to a primary or secondary amine. The dye is a superior alternative to 5(6)-SFX because of its exceptional phototostability and fluorescence insensitivity to pH between 4-9.Properties: Ex / Em (MeOH) = 502/524 nm; Orange red solid soluble in DMSO or DMF; Store at -20°C and protect from light; C 31 H 28 N 4 O 8; MW: 584.58
Catalog No. 50-196-4798
Edge