Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Diacetoxyindole 98.0+%, TCI America™
SDP

Catalog No. D23171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23171G 1 g
D23175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23171G Supplier TCI America Supplier No. D23171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15069-79-1
Molecular Formula C12H11NO4
Molecular Weight (g/mol) 233.22
MDL Number MFCD00152051
InChI Key NTOLUQGMBCPVOZ-UHFFFAOYSA-N
PubChem CID 3805883
IUPAC Name 5-(acetyloxy)-1H-indol-6-yl acetate
SMILES CC(=O)OC1=C(OC(C)=O)C=C2C=CNC2=C1

Specifications

Melting Point 138°C
Quantity 1 g
Formula Weight 233.22
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Diacetoxyindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.