Learn More
Medchemexpress LLC 5,6-Diamino-1,10-phenanthroline | 168646-54-6 | 210.23 | 1 G
SDP
Catalog No. 5000446937
Encompass_Preferred
5,6-Diamino-1,10-phenanthroline is a compound that can be used as ligands for metal ions for drug synthesis. It is intended for research use only.
  • Can be used as ligands for metal ions
  • Suitable for drug synthesis
  • For research use only
Catalog No. 50-004-46937 Supplier Medchemexpress LLC Supplier No. HYW0045641G
5,6-Diamino-1,10-phenanthroline is a compound that can be used as ligands for metal ions for drug synthesis. It is intended for research use only.
  • Can be used as ligands for metal ions
  • Suitable for drug synthesis
  • For research use only