Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,6-Diamino-1,10-phenanthroline | 168646-54-6 | 210.23 | 100 MG
SDP

Catalog No. 5000448806
Encompass_Preferred

5,6-Diamino-1,10-phenanthroline can be used as ligands for metal ions for drug synthesis.

  • Purity of 97.13%
  • Solid appearance
  • Yellow to orange color
  • Molecular weight of 210.23
  • Chemical formula C12H10N4
  • Ships at room temperature in continental US (may vary elsewhere)
  • Storage at 4°C, protected from light and under nitrogen
  • In solvent: store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Catalog No. 50-004-48806 Supplier Medchemexpress LLC Supplier No. HYW004564100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,6-Diamino-1,10-phenanthroline can be used as ligands for metal ions for drug synthesis.

  • Purity of 97.13%
  • Solid appearance
  • Yellow to orange color
  • Molecular weight of 210.23
  • Chemical formula C12H10N4
  • Ships at room temperature in continental US (may vary elsewhere)
  • Storage at 4°C, protected from light and under nitrogen
  • In solvent: store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.