Learn More
Medchemexpress LLC 5,6-Diamino-1,10-phenanthroline | 168646-54-6 | 210.23 | 500 MG

Supplier: Medchemexpress LLC HYW004564500MG
5,6-Diamino-1,10-phenanthroline can be used as ligands for metal ions for drug synthesis. It is intended for research use only, and not sold to patients.
- Used as ligands for metal ions for drug synthesis
- Appearance: solid
- Color: yellow to orange
- Solid storage: 4°C, protected from light, stored under nitrogen
- Solvent storage: -80°C for 6 months, or -20°C for 1 month (protected from light, stored under nitrogen)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.