Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Diamino-2,3-dicyanopyrazine 98.0+%, TCI America™
SDP

Catalog No. D227010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D227010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D227010G Supplier TCI America Supplier No. D227010G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36023-58-2
Molecular Formula C6H4N6
Molecular Weight (g/mol) 160.14
MDL Number MFCD00191418
InChI Key FTHBTDDIVWLRLP-UHFFFAOYSA-N
Synonym 5,6-Diamino-2,3-pyrazinedicarbonitrile
PubChem CID 4120764
IUPAC Name 5,6-diaminopyrazine-2,3-dicarbonitrile
SMILES C(#N)C1=C(N=C(C(=N1)N)N)C#N

Specifications

Melting Point 332°C
Color Yellow
Quantity 10 g
UN Number 2811
Formula Weight 160.14
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Diamino-2,3-dicyanopyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.