Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dichlorobenzimidazole 98.0+%, TCI America™
SDP

Catalog No. D51461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51461G 1 g
D51465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51461G Supplier TCI America Supplier No. D51461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6478-73-5
Molecular Formula C7H4Cl2N2
Molecular Weight (g/mol) 187.02
MDL Number MFCD00211325
InChI Key IPRDZAMUYMOJTA-UHFFFAOYSA-N
PubChem CID 331775
IUPAC Name 5,6-dichloro-1H-1,3-benzodiazole
SMILES ClC1=C(Cl)C=C2N=CNC2=C1

Specifications

Melting Point 210°C
Quantity 1 g
Formula Weight 187.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichlorobenzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.