Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Difluorosalicylaldehyde, 98%, Thermo Scientific™

Catalog No. AAH3214603
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3214603 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3214603 Supplier Thermo Scientific Chemicals Supplier No. H3214603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 187543-89-1
Molecular Formula C7H4F2O2
Quantity 1g
MDL Number MFCD08275282
Synonym 5,6-difluorosalicylaldehyde, benzaldehyde, 2,3-difluoro-6-hydroxy-9ci, 3,4-difluoro-2-formylphenol, benzaldehyde,2,3-difluoro-6-hydroxy, benzaldehyde, 2,3-difluoro-6-hydroxy
InChI Key HCXZOHFGXHDGRF-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1O)C=O)F)F
IUPAC Name 2,3-difluoro-6-hydroxybenzaldehyde
Molecular Weight (g/mol) 158.104
PubChem CID 21221031
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material 5,6-Difluorosalicylaldehyde
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.