Learn More
Cayman Chemical 5 6-DIHET lacton 100UG
SDP
Catalog No. 502893182
Encompass_Preferred
A lactone form of (±)5(6)-DiHET; induces vasodilation of isolated canine coronary arterioles, with 41 and 100% inhibition occurring at 0.01 and 100 pM, respectively; induces vasodilation in isolated human microvessels and increases intracellular calcium levels in a dose-dependent manner
Catalog No. 50-289-3182 Supplier Cayman Chemical Supplier No. 51212100 UG
A lactone form of (±)5(6)-DiHET; induces vasodilation of isolated canine coronary arterioles, with 41 and 100% inhibition occurring at 0.01 and 100 pM, respectively; induces vasodilation in isolated human microvessels and increases intracellular calcium levels in a dose-dependent manner