Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dihydro-1,4-dithiin-2,3-dicarboxylic Anhydride 98.0+%, TCI America™
SDP

Catalog No. D25751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25751G 1 g
D25755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25751G Supplier TCI America Supplier No. D25751G

Chemical Identifiers

CAS 10489-75-5
Molecular Formula C6H4O3S2
Molecular Weight (g/mol) 188.215
MDL Number MFCD00223990
InChI Key MXSSHXZXAAXCOW-UHFFFAOYSA-N
Synonym 2,3-Dihydro-1,4-dithiino[2,3-c]furan-5,7-dione
PubChem CID 82669
IUPAC Name 2,3-dihydro-[1,4]dithiino[2,3-c]furan-5,7-dione
SMILES C1CSC2=C(S1)C(=O)OC2=O

Specifications

Melting Point 112°C
Quantity 1 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dihydro-1,4-dithiin-2,3-dicarboxylic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.