Learn More
Medchemexpress LLC 5,6-dihydro-5-fluorouracil | 696-06-0 | MFCD00871150 | 99.9% | 132.09 g·mol⁻¹ | C4H5FN2O2 | 5 MG
SDP
Catalog No. 5000222441
Encompass_Preferred
5,6-Dihydro-5-fluorouracil (5-DHFU) is the active metabolite of the chemotherapy prodrug 5-fluorouracil. Supplied as a white to off-white solid for research use, it is used in metabolic profiling, enzyme assays, and studies of drug metabolism and pharmacokinetics. The material is suitable as a reference standard in biochemical and pharmacological investigations.
  • High purity suitable for analytical and preparative studies.
  • Available in small milligram quantities for early-stage research.
  • Solid form facilitates weighing, storage, and transport.
  • Documented chemical identifiers support traceability.
  • Commonly used as a reference in metabolic and enzymatic assays.
Catalog No. 50-002-22441 Supplier Medchemexpress LLC Supplier No. HY1341605MG
5,6-Dihydro-5-fluorouracil (5-DHFU) is the active metabolite of the chemotherapy prodrug 5-fluorouracil. Supplied as a white to off-white solid for research use, it is used in metabolic profiling, enzyme assays, and studies of drug metabolism and pharmacokinetics. The material is suitable as a reference standard in biochemical and pharmacological investigations.
  • High purity suitable for analytical and preparative studies.
  • Available in small milligram quantities for early-stage research.
  • Solid form facilitates weighing, storage, and transport.
  • Documented chemical identifiers support traceability.
  • Commonly used as a reference in metabolic and enzymatic assays.