Learn More
Medchemexpress LLC 5,6-dihydro-5-fluorouracil | 696-06-0 | MFCD00871150 | 99.9% | C4H5FN2O2 | 10MG
SDP
Catalog No. 5000259469
Encompass_Preferred
5,6-Dihydro-5-fluorouracil is a fluorinated dihydropyrimidine metabolite used as a research reagent and analytical standard in biochemical and medicinal chemistry studies. It is supplied as a solid, high-purity material suitable for in vitro assays, metabolite profiling, and method development.
  • High purity of 99.9% for analytical reliability.
  • Solid, white to off-white appearance for easy handling.
  • Molecular formula C4H5FN2O2 and molecular weight 132.09 g/mol.
  • Available in small pack sizes including 10 mg for scarce-sample experiments.
  • Stable as a powder at -20°C for long-term storage.
  • Includes structural identifier (SMILES) for informatics and modeling.
Catalog No. 50-002-59469 Supplier Medchemexpress LLC Supplier No. HY13416010MG
5,6-Dihydro-5-fluorouracil is a fluorinated dihydropyrimidine metabolite used as a research reagent and analytical standard in biochemical and medicinal chemistry studies. It is supplied as a solid, high-purity material suitable for in vitro assays, metabolite profiling, and method development.
  • High purity of 99.9% for analytical reliability.
  • Solid, white to off-white appearance for easy handling.
  • Molecular formula C4H5FN2O2 and molecular weight 132.09 g/mol.
  • Available in small pack sizes including 10 mg for scarce-sample experiments.
  • Stable as a powder at -20°C for long-term storage.
  • Includes structural identifier (SMILES) for informatics and modeling.