Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,6-Dihydro-5-methyluracil-d6 | 334473-42-6 | 1 MG
SDP

Catalog No. 5000420204
Encompass_Preferred

5,6-Dihydro-5-methyluracil-d6 is the deuterium-labeled version of 5,6-Dihydro-5-methyluracil (Dihydrothymine). Dihydrothymine is an intermediate breakdown product of thymine, found in animals and plants, and can be toxic at abnormally high levels. This compound is intended for research use only and is not sold to patients.

  • Application as a tracer
  • Internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs
  • High purity (99%)
  • Appearance: solid, white to off-white in color
  • Molecular formula: C5H2D6N2O2
  • Molecular weight: 134.17

Catalog No. 50-004-20204 Supplier Medchemexpress LLC Supplier No. HYN6787S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,6-Dihydro-5-methyluracil-d6 is the deuterium-labeled version of 5,6-Dihydro-5-methyluracil (Dihydrothymine). Dihydrothymine is an intermediate breakdown product of thymine, found in animals and plants, and can be toxic at abnormally high levels. This compound is intended for research use only and is not sold to patients.

  • Application as a tracer
  • Internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs
  • High purity (99%)
  • Appearance: solid, white to off-white in color
  • Molecular formula: C5H2D6N2O2
  • Molecular weight: 134.17

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.