Learn More
Apexbio Technology LLC 5,6-Dihydro-UTP(Synonyms: 5,6-Dihydrouridine-5'-triphosphate, DHU-TP, 5,6-DHUTP, Dihydrouridine triphosphate), 10ul (100 mM).
SDP
Catalog No. 5000061278
Encompass_Preferred
5 6-Dihydro-UTP is a base-modified ribonucleoside triphosphate utilized in biochemical research Acting primarily as an acceptor substrate during enzymatic synthesis of dinucleoside polyphosphates this compound participates in phosphorylation reactions catalyzed by nucleotide-utilizing enzymes Due to its altered nucleobase structure 5 6-Dihydro-UTP serves as a strategic molecular probe in studies examining nucleotide synthesis pathways enzyme specificity and nucleic acid biochemistry
Catalog No. 50-000-61278 Supplier Apexbio Technology LLC Supplier No. B79870.01
5 6-Dihydro-UTP is a base-modified ribonucleoside triphosphate utilized in biochemical research Acting primarily as an acceptor substrate during enzymatic synthesis of dinucleoside polyphosphates this compound participates in phosphorylation reactions catalyzed by nucleotide-utilizing enzymes Due to its altered nucleobase structure 5 6-Dihydro-UTP serves as a strategic molecular probe in studies examining nucleotide synthesis pathways enzyme specificity and nucleic acid biochemistry