Learn More
Medchemexpress LLC HY-W012926 100mg Medchemexpress, 5,6-Dihydrouracil CAS:504-07-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW012926100MG
Encompass_Preferred
Medchemexpress, HY-W012926 100mg 5,6-Dihydrouracil CAS:504-07-4 5,6-Dihydrouracil is an intermediate breakdown product of uracil. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4574