Learn More
Cayman Chemical 5,6-Dihydrouridine, Purity: ≥95%, CAS: 5627-05-4, 250 mg
SDP
Catalog No. 502949495
Encompass_Preferred
A pyrimidine nucleoside and derivative of uridine; present in tRNA from bacteria, eukaryotes, and some archaea; inhibits E. coli cytidine deaminase in a cell-free assay (Ki = 3.4 µM); serum levels are increased in patients with prostate cancer and positively correlated with lethality
Catalog No. 50-294-9495 Supplier Cayman Chemical Supplier No. 32523250 MG
A pyrimidine nucleoside and derivative of uridine; present in tRNA from bacteria, eukaryotes, and some archaea; inhibits E. coli cytidine deaminase in a cell-free assay (Ki = 3.4 µM); serum levels are increased in patients with prostate cancer and positively correlated with lethality