Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,6-dihydroxyindole | 3131-52-0 | MFCD00798933 | 99.3% | 149.15 g/mol | C8H7NO2 | 5 G
SDP

Catalog No. 5000300786
Encompass_Preferred

5,6-Dihydroxyindole (CAS 3131-52-0) is an indole derivative and a precursor in eumelanin biosynthesis supplied as a solid research reagent. It is intended for biochemical studies, organic synthesis, and materials research requiring dihydroxyindole functionality.

  • High purity (≈99.3%) suitable for research applications.
  • Available in multiple pack sizes, including a 5 g pack for preparative work.
  • Off-white to light brown solid, convenient for weighing and handling.
  • Storage recommendations: protect from light, store under nitrogen at -20°C for long-term stability.
  • Molecular weight 149.15 g/mol; formula C8H7NO2; CAS 3131-52-0.

Catalog No. 50-003-00786 Supplier Medchemexpress LLC Supplier No. HYW0180255G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,6-Dihydroxyindole (CAS 3131-52-0) is an indole derivative and a precursor in eumelanin biosynthesis supplied as a solid research reagent. It is intended for biochemical studies, organic synthesis, and materials research requiring dihydroxyindole functionality.

  • High purity (≈99.3%) suitable for research applications.
  • Available in multiple pack sizes, including a 5 g pack for preparative work.
  • Off-white to light brown solid, convenient for weighing and handling.
  • Storage recommendations: protect from light, store under nitrogen at -20°C for long-term stability.
  • Molecular weight 149.15 g/mol; formula C8H7NO2; CAS 3131-52-0.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.