Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dimethoxy-2-(4-piperidylmethyl)-1-indanone Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D39975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39975G 5 g
D39971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39975G Supplier TCI America Supplier No. D39975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120013-39-0
Molecular Formula C17H24ClNO3
Molecular Weight (g/mol) 325.83
MDL Number MFCD09033179
InChI Key WOZXDQQCMZIQEG-UHFFFAOYNA-N
PubChem CID 14553606
IUPAC Name 5,6-dimethoxy-2-[(piperidin-4-yl)methyl]-2,3-dihydro-1H-inden-1-one hydrochloride
SMILES Cl.COC1=C(OC)C=C2C(=O)C(CC3CCNCC3)CC2=C1

Specifications

Melting Point 250°C
Color Yellow
Quantity 5 g
Formula Weight 325.83
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dimethoxy-2-(4-piperidylmethyl)-1-indanone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.