Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dimethyl-1,10-phenanthroline 98.0+%, TCI America™
SDP

Catalog No. D15591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15591G 1 g
D1559100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15591G Supplier TCI America Supplier No. D15591G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3002-81-1
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD00004983
InChI Key BRPQDJPJBCQFSR-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 5,6-dimethyl, acmc-1afjl, brpqdjpjbcqfsr-uhfffaoysa, 5,6-dimethylpyridino 3,2-h quinoline, 5,6-dimethyl-1,10-phenanthroline
PubChem CID 76357
IUPAC Name 5,6-dimethyl-1,10-phenanthroline
SMILES CC1=C2C=CC=NC2=C3C(=C1C)C=CC=N3

Specifications

Color Yellow
Quantity 1 g
Formula Weight 208.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dimethyl-1,10-phenanthroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.