Learn More
Cayman Chemical 5,6-dimethyl-2-Thiouracil, Purity: ≥95%, CAS: 28456-54-4, 1 g
SDP
Catalog No. 502990228
Encompass_Preferred
A heterocyclic building block; has been used in the synthesis of anti-HIV-1 pyrimidinones; has been used as an internal standard for the quantification of thyreostats in bovine plasma
Catalog No. 50-299-0228 Supplier Cayman Chemical Supplier No. 298211 G
A heterocyclic building block; has been used in the synthesis of anti-HIV-1 pyrimidinones; has been used as an internal standard for the quantification of thyreostats in bovine plasma