Learn More
Cayman Chemical 5,6-dimethyl-2-Thiouracil, Purity: ≥95%, CAS: 28456-54-4, 5 g
SDP
Catalog No. 502988449
Encompass_Preferred
A heterocyclic building block; has been used in the synthesis of anti-HIV-1 pyrimidinones; has been used as an internal standard for the quantification of thyreostats in bovine plasma
Catalog No. 50-298-8449 Supplier Cayman Chemical Supplier No. 298215 G
A heterocyclic building block; has been used in the synthesis of anti-HIV-1 pyrimidinones; has been used as an internal standard for the quantification of thyreostats in bovine plasma