Learn More
Cayman Chemical 5 6-epoxy Cholestnol 100mg
SDP
Catalog No. 502936075
Encompass_Preferred
An oxidative metabolite of cholesterol; formed via radical and non-radical oxidation of cholesterol at the 5,6-double bond; induces release of LDH and apoptosis in macrophage-differentiated U937 cells; has been found in human fatty streaks and advanced atherosclerotic lesions
Catalog No. 50-293-6075 Supplier Cayman Chemical Supplier No. 25603100 MG
An oxidative metabolite of cholesterol; formed via radical and non-radical oxidation of cholesterol at the 5,6-double bond; induces release of LDH and apoptosis in macrophage-differentiated U937 cells; has been found in human fatty streaks and advanced atherosclerotic lesions