Learn More
Cayman Chemical 5β,6β-epoxy Cholestanol, Purity: ≥95%, CAS: 4025-59-6, 50 mg
SDP
Catalog No. 502936413
Encompass_Preferred
An oxidative metabolite of cholesterol; formed via radical and non-radical oxidation of cholesterol at the 5,6-double bond; induces release of LDH and apoptosis in macrophage-differentiated U937 cells; has been found in human fatty streaks and advanced atherosclerotic lesions
Catalog No. 50-293-6413 Supplier Cayman Chemical Supplier No. 2560350 MG
An oxidative metabolite of cholesterol; formed via radical and non-radical oxidation of cholesterol at the 5,6-double bond; induces release of LDH and apoptosis in macrophage-differentiated U937 cells; has been found in human fatty streaks and advanced atherosclerotic lesions