Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 5(6)-FAM, SE (5-(and-6)-Carboxyfluorescein, Succinimidyl Ester), mixed isomers

Catalog No. C1311
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1311 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1311 Supplier Invitrogen™ Supplier No. C1311
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

5(6)-FAM, SE (5-(and-6)-Carboxyfluorescein, Succinimidyl Ester), mixed isomers

Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Product Type 5(6)-FAM SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.