Learn More
Medchemexpress LLC 5-[6-methyl-5-[(1S,2R)-2-propan-2-ylcyclopropyl]pyridazin-3-yl]-1H-pyrimidine-2,4-dione | 2375815-63-5 | 99.9% | 286.33 g/mol | C15H18N4O2 | 5 MG
SDP
Catalog No. 5000222450
Encompass_Preferred
CD73-IN-3 is a small-molecule CD73 inhibitor intended for preclinical cancer research and cell-based assays. It is supplied as a high-purity solid and has been characterized in human cell assays for potency and stability.
  • Potent CD73 inhibition with IC50 = 7.3 nM in Calu6 human cell assay.
  • High purity (99.89%) suitable for research applications.
  • Solid, white to off-white physical form for easy handling and storage.
  • Stable under recommended storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Molecular formula C15H18N4O2 and molecular weight 286.33 g/mol for formulation planning.
Catalog No. 50-002-22450 Supplier Medchemexpress LLC Supplier No. HY1372465MG
CD73-IN-3 is a small-molecule CD73 inhibitor intended for preclinical cancer research and cell-based assays. It is supplied as a high-purity solid and has been characterized in human cell assays for potency and stability.
  • Potent CD73 inhibition with IC50 = 7.3 nM in Calu6 human cell assay.
  • High purity (99.89%) suitable for research applications.
  • Solid, white to off-white physical form for easy handling and storage.
  • Stable under recommended storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Molecular formula C15H18N4O2 and molecular weight 286.33 g/mol for formulation planning.