Learn More
AdipoGen 5(6)-ROX N-succinimidyl ester,Chemical
SDP
Supplier:  AdipoGen CDXC0013M010
Encompass_Preferred
5(6)-ROX N-succinimidyl ester, AdipoGen Life Sciences. Chemical. CAS: 114616-32-9. Formula: C37H33N3O7. MW: 631.67. Synthetic. 5-(and-6)-Carboxy-X-rhodamine, succinimidyl ester is an amine-reactive long wavelength rhodamine dye. Derivatives made with the amine-reactive carboxy-X-rhodamine (ROX), succinimidyl ester are widely used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lissamine rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red¿ conjugates. 114616-32-9
Catalog No. 50-117-7267