Learn More
Aat Bioquest 5(6)-TAMRA Maleimide [Tetramethylrhodamine-5-(and-6)-maleimide] *Mixed isomers*
SDP
Catalog No. 501952880
Encompass_Preferred
Tetramethylrhodamine-5-(and-6)-maleimide is an excellent reagent for preparing orange fluorescent bioconjugates through thiol modifications of peptides, nucleotides, nucleic acids and proteins.
Catalog No. 50-195-2880 Supplier Aat Bioquest Supplier No. 412
Tetramethylrhodamine-5-(and-6)-maleimide is an excellent reagent for preparing orange fluorescent bioconjugates through thiol modifications of peptides, nucleotides, nucleic acids and proteins.