Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,6-trans-ergocalciferol | 51744-66-2 | 98.8% | 396.65 | C28H44O | 10 MG
SDP

Supplier:  Medchemexpress LLC HY76542A10MG

Encompass_Preferred

5,6-trans-Vitamin D2 is the 5,6-trans isomer of vitamin D2 supplied as an analytical standard for research and analytical applications. It is provided as an off-white to light yellow solid with high purity and defined chemical identifiers for use as a reference or control in analytical workflows.

  • High purity suitable for analytical standards (98.77%).
  • Available in small quantities for analytical use (5 MG, 10 MG, 25 MG, 50 MG, 100 MG).
  • Chemical formula C28H44O, molecular weight 396.65, CAS 51744-66-2.
  • Appearance off-white to light yellow solid.
  • Storage: protect from light, store under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month; solid: -20°C.
  • Includes structural identifiers for unambiguous identification in analytical methods.

Catalog No. 50-002-98185


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.