Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,6-trans-Vitamin D2 | 51744-66-2 | 97.0% | C28H44O | 100 MG
SDP

Catalog No. 5000434141
Encompass_Preferred

5,6-trans-Vitamin D2 is an isomer of Vitamin D2. It is supplied as an off-white to light yellow solid with a molecular weight of 396.65. This product is for research use only and not intended for patient use.

  • Isomer of vitamin D2
  • For research use only
  • Molecular weight of 396.65
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO (25 mg/mL)
  • In vivo dissolution protocols available

Catalog No. 50-004-34141 Supplier Medchemexpress LLC Supplier No. HY76542A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,6-trans-Vitamin D2 is an isomer of Vitamin D2. It is supplied as an off-white to light yellow solid with a molecular weight of 396.65. This product is for research use only and not intended for patient use.

  • Isomer of vitamin D2
  • For research use only
  • Molecular weight of 396.65
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO (25 mg/mL)
  • In vivo dissolution protocols available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.