Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,6-trans-vitamin D2 | 51744-66-2 | 97.0% | C28H44O | 50 MG
SDP

Catalog No. 5000434107
Encompass_Preferred

5,6-trans-Vitamin D2 is an isomer of Vitamin D2. It is for research use only. This product is an off-white to light yellow solid.

  • Isomer of Vitamin D2
  • Off-white to light yellow solid
  • For research use only
  • Solubility in DMSO: 25 mg/mL (63.03 mM)
  • Recommended storage for solid: -20°C, protect from light, stored under nitrogen
  • Recommended storage in solvent: -80°C for 6 months, or -20°C for 1 month (protect from light, stored under nitrogen)

Catalog No. 50-004-34107 Supplier Medchemexpress LLC Supplier No. HY76542A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,6-trans-Vitamin D2 is an isomer of Vitamin D2. It is for research use only. This product is an off-white to light yellow solid.

  • Isomer of Vitamin D2
  • Off-white to light yellow solid
  • For research use only
  • Solubility in DMSO: 25 mg/mL (63.03 mM)
  • Recommended storage for solid: -20°C, protect from light, stored under nitrogen
  • Recommended storage in solvent: -80°C for 6 months, or -20°C for 1 month (protect from light, stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.