Learn More
Enzo Life Sciences 5(6)-TRITC (ultra pure) (10mg). CAS: 95197-95-8
SDP
Supplier:  Enzo Life Sciences ENZ52452
Encompass_Preferred
5(6)-TRITC is an amino-reactive labeling reagent that is widely used in preparing bioconjugates of proteins and nucleic acids. The resultant conjugates have similar spectral properties to those prepared from 5(6)-TAMRA, SE with improved stability. TRITC is widely used and has a multitude of applications beyond protein, peptide and nucleotide labeling For example, it can serve as an acceptor for FRET (Fluorescence Resonance Energy Transfer) based detection systems. FITC is recommended as a donor molecule in these FRET-based applications. Wavelength Maxima: Excitation 543nm, Emission 571nm Alternative name: Tetramethylrhodamine-5(6)-isothiocyanate. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2641