Learn More
Cayman Chemical 5 6-trns-VItamIn D3 10mg
SDP
Catalog No. 502990020
Encompass_Preferred
An isomer of vitamin D3; increases tibia ash weight and bone mineralization in vitamin D-deficient chicks at 1, 3, and 10 UG/day; induces intestinal calcium transport and bone calcium mobilization in anephric rats fed a low-calcium and vitamin D-deficient diet at 25 UG/animal
Catalog No. 50-299-0020 Supplier Cayman Chemical Supplier No. 3122510 MG
Add to Cart
Add to Cart
An isomer of vitamin D3; increases tibia ash weight and bone mineralization in vitamin D-deficient chicks at 1, 3, and 10 UG/day; induces intestinal calcium transport and bone calcium mobilization in anephric rats fed a low-calcium and vitamin D-deficient diet at 25 UG/animal