Learn More
Cayman Chemical 5 6-trns-VItamIn D3 1mg
SDP
Supplier:  Cayman Chemical 312251 MG
Encompass_Preferred
An isomer of vitamin D3; increases tibia ash weight and bone mineralization in vitamin D-deficient chicks at 1, 3, and 10 UG/day; induces intestinal calcium transport and bone calcium mobilization in anephric rats fed a low-calcium and vitamin D-deficient diet at 25 UG/animal
Catalog No. 50-298-8418