Learn More
Cayman Chemical 5,6-trans-Vitamin D3, Purity: ≥98%, CAS: 22350-41-0, 5 mg
SDP
Catalog No. 502990022
Encompass_Preferred
An isomer of vitamin D3; increases tibia ash weight and bone mineralization in vitamin D-deficient chicks at 1, 3, and 10 µg/day; induces intestinal calcium transport and bone calcium mobilization in anephric rats fed a low-calcium and vitamin D-deficient diet at 25 µg/animal
Catalog No. 50-299-0022 Supplier Cayman Chemical Supplier No. 312255 MG
An isomer of vitamin D3; increases tibia ash weight and bone mineralization in vitamin D-deficient chicks at 1, 3, and 10 µg/day; induces intestinal calcium transport and bone calcium mobilization in anephric rats fed a low-calcium and vitamin D-deficient diet at 25 µg/animal