Learn More
Cayman Chemical 5 6CarboxyfluorSceIn dIAC 50mg
SDP
Catalog No. 502984547
Encompass_Preferred
A fluorescent probe for staining cells with intact membranes (excitation/emission at 494/521 nm)
Catalog No. 50-298-4547 Supplier Cayman Chemical Supplier No. 1958250 MG
A fluorescent probe for staining cells with intact membranes (excitation/emission at 494/521 nm)