Learn More
Cayman Chemical 5 6dehydro ArachIdonIc ACD
SDP
Catalog No. 502893067
Encompass_Preferred
An analog of AA with the triple bond in the 5,6 position; inhibitor of 5-LO (Ki = 10-15 μM)
Catalog No. 50-289-3067 Supplier Cayman Chemical Supplier No. 90020100 UG
An analog of AA with the triple bond in the 5,6 position; inhibitor of 5-LO (Ki = 10-15 μM)