Learn More
Cayman Chemical 5,6-dihydro-5-Fluorouracil, Purity: ≥95%, CAS: 696-06-0, 10 mg
SDP
Catalog No. 502983776
Encompass_Preferred
An active metabolite of 5-fluorouracil; is cytotoxic to HaCaT cells (IC50 = 13.5 µM); reduces tumor growth in combination with 5-fluorouracil and eniluracil in a rat model of colon carcinoma at 90 mg/kg per week
Catalog No. 50-298-3776 Supplier Cayman Chemical Supplier No. 2097710 MG
An active metabolite of 5-fluorouracil; is cytotoxic to HaCaT cells (IC50 = 13.5 µM); reduces tumor growth in combination with 5-fluorouracil and eniluracil in a rat model of colon carcinoma at 90 mg/kg per week