Learn More
Cayman Chemical 5 6EETd11 GC MS Standards
SDP
Catalog No. 502889169
Encompass_Preferred
An internal standard for the quantification of 5(6)-EET by GC- or LC-MS
Catalog No. 50-288-9169 Supplier Cayman Chemical Supplier No. 1000998410 UG
An internal standard for the quantification of 5(6)-EET by GC- or LC-MS