Learn More
Cayman Chemical 5 6EETd11 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000998425 UG
Encompass_Preferred
An internal standard for the quantification of 5(6)-EET by GC- or LC-MS
Catalog No. 50-288-8479