Learn More
Cayman Chemical 5 6EETd11 methyl ester 100ug
SDP
Catalog No. 502988131
Encompass_Preferred
An internal standard for the quantification of 5(6)-EET methyl ester by GC- or LC-MS
Catalog No. 50-298-8131 Supplier Cayman Chemical Supplier No. 29382100 UG
An internal standard for the quantification of 5(6)-EET methyl ester by GC- or LC-MS