Learn More
Chemglass Life Sciences 5 and 6mm Bore, 16/35 Plug Size, 12mm OD Sidearm
SDP
Catalog No. 501531996
Encompass_Preferred
5 and 6mm Bore, 16/35 Plug Size, 12mm OD Sidearm. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.
Catalog No. 50-153-1996 Supplier Chemglass Life Sciences Supplier No. CGB42607
5 and 6mm Bore, 16/35 Plug Size, 12mm OD Sidearm. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.