Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 5mm & 6mm Bore, 16/35 Barrel Size, 12mm OD Sidearms
SDP
Catalog No. CG42607
Encompass_Preferred
Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 5mm & 6mm Bore, 16/35 Barrel Size, 12mm OD Sidearms. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.
Catalog No. CG42607 Supplier Chemglass Life Sciences Supplier No. CG42607
Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 5mm & 6mm Bore, 16/35 Barrel Size, 12mm OD Sidearms. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.