Learn More
Cayman Chemical 5,7,3',4',5'-Pentamethoxyflavone, Purity: ≥98%, CAS: 53350-26-8, 50 mg
SDP
Catalog No. 502942037
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits pancreatic lipase in a cell-free assay; reduces lipid accumulation in 3T3-L1 adipocytes; inhibits transactivation of Nrf2 in a reporter assay using HEK293T cells at 10, 25, or 50 µM; sensitizes cisplatin-resistant A549 NSCLC cells to cisplatin; inhibits LPS-induced NO production in RAW 264.7 macrophages and GES-1 cells; decreases blood glucose levels and renal fibrosis in a rat model of STZ-induced diabetes at 5 mg/kg
Catalog No. 50-294-2037 Supplier Cayman Chemical Supplier No. 3982050 MG
A flavonoid with diverse biological activities; inhibits pancreatic lipase in a cell-free assay; reduces lipid accumulation in 3T3-L1 adipocytes; inhibits transactivation of Nrf2 in a reporter assay using HEK293T cells at 10, 25, or 50 µM; sensitizes cisplatin-resistant A549 NSCLC cells to cisplatin; inhibits LPS-induced NO production in RAW 264.7 macrophages and GES-1 cells; decreases blood glucose levels and renal fibrosis in a rat model of STZ-induced diabetes at 5 mg/kg