Learn More
TARGETMOL CHEMICALS INC Research Chemical 25mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T822925MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5,7,3',4'-Tetramethoxyflavone (TMF), a prominent polymethoxyflavone (PMF) derived from M. exotica, displays a range of notable bioactivities. These include anti-malarial, anti-fungal, anti-mycobacterial, and anti-inflammatory properties. Moreover, TMF specifically targets (beta)-catenin signaling to exhibit chondroprotective activity. purity: 99%
Catalog No. 50-303-8157