Learn More
Medchemexpress LLC 5,7,3',4'-Tetramethoxyflavone | 855-97-0 | 10 MG
SDP
Catalog No. 5000419605
Encompass_Preferred
5,7,3',4'-Tetramethoxyflavone is an orally active polymethoxyflavone (PMF) isolated from M. exotica. It possesses various bioactivities, including anti-fungal, anti-malarial, anti-mycobacterial, and anti-inflammatory activities. It also exhibits chondroprotective activity by targeting β-catenin signaling.
  • Possesses anti-fungal, anti-malarial, anti-mycobacterial, and anti-inflammatory properties.
  • Exhibits chondroprotective activity by targeting β-catenin signaling.
  • Protects chondrocytes from ER stress-induced apoptosis.
  • Decreases inflammatory cytokines.
  • Inhibits chondrocyte hypertrophy.
Catalog No. 50-004-19605 Supplier Medchemexpress LLC Supplier No. HYN703010MG
5,7,3',4'-Tetramethoxyflavone is an orally active polymethoxyflavone (PMF) isolated from M. exotica. It possesses various bioactivities, including anti-fungal, anti-malarial, anti-mycobacterial, and anti-inflammatory activities. It also exhibits chondroprotective activity by targeting β-catenin signaling.
  • Possesses anti-fungal, anti-malarial, anti-mycobacterial, and anti-inflammatory properties.
  • Exhibits chondroprotective activity by targeting β-catenin signaling.
  • Protects chondrocytes from ER stress-induced apoptosis.
  • Decreases inflammatory cytokines.
  • Inhibits chondrocyte hypertrophy.