Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,7,4'-Trihydroxy-8-Methylflavanone | 916917-28-7 | 1 MG
SDP

Catalog No. 5000418695
Encompass_Preferred

5,7,4'-Trihydroxy-8-Methylflavanone (Compound 2) is a flavanone isolated from Qualea grandiflora. It is intended for research use only.

  • Solid appearance
  • White to off-white color
  • Classified as flavonoids, flavanones, phenols, and polyphenols
  • Initial source from plants, Ericaceae, Rhododendron dauricum Linn.
  • Purity of 99.94%
  • Molecular weight of 286.28
  • Molecular formula C16H14O5

Catalog No. 50-004-18695 Supplier Medchemexpress LLC Supplier No. HYN22101MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,7,4'-Trihydroxy-8-Methylflavanone (Compound 2) is a flavanone isolated from Qualea grandiflora. It is intended for research use only.

  • Solid appearance
  • White to off-white color
  • Classified as flavonoids, flavanones, phenols, and polyphenols
  • Initial source from plants, Ericaceae, Rhododendron dauricum Linn.
  • Purity of 99.94%
  • Molecular weight of 286.28
  • Molecular formula C16H14O5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.