Learn More
Medchemexpress LLC 5,7,4'-Trihydroxy-8-Methylflavanone | 916917-28-7 | 5 MG
SDP
Catalog No. 5000420760
Encompass_Preferred
5,7,4'-Trihydroxy-8-Methylflavanone, also known as 8-Methyl-naringenine, is a high-purity flavanone compound (99.94%) primarily isolated from *Qualea grandiflora*. This product is intended strictly for research applications and is not for patient use. It is a solid, white to off-white substance with a molecular weight of 286.28 and a chemical formula of C16H14O5. Classified within flavonoids and flavonones, as well as phenols and polyphenols, it can also be sourced from plants like *Ericaceae* and *Rhododendron dauricum Linn.*.
  • High purity compound (99.94%)
  • Isolated from natural sources such as *Qualea grandiflora*
  • Suitable for various research applications
  • Identified as a flavanone, phenol, and polyphenol
  • Stable for storage at 4°C in sealed conditions, away from moisture
  • Can be stored in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-20760 Supplier Medchemexpress LLC Supplier No. HYN22105MG
5,7,4'-Trihydroxy-8-Methylflavanone, also known as 8-Methyl-naringenine, is a high-purity flavanone compound (99.94%) primarily isolated from *Qualea grandiflora*. This product is intended strictly for research applications and is not for patient use. It is a solid, white to off-white substance with a molecular weight of 286.28 and a chemical formula of C16H14O5. Classified within flavonoids and flavonones, as well as phenols and polyphenols, it can also be sourced from plants like *Ericaceae* and *Rhododendron dauricum Linn.*.
  • High purity compound (99.94%)
  • Isolated from natural sources such as *Qualea grandiflora*
  • Suitable for various research applications
  • Identified as a flavanone, phenol, and polyphenol
  • Stable for storage at 4°C in sealed conditions, away from moisture
  • Can be stored in solvent at -80°C for 6 months or -20°C for 1 month