Learn More
TARGETMOL CHEMICALS INC 5 7 4 -Trimethoxyflavone 10MG
SDP
Catalog No. 502879153
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 5,7,4'-Trimethoxyflavone (4',5,7-Trimethoxyflavone) is isolated from Kaempferia parviflora (KP) which is a medicinal plant from Thailand. Purity 97.53%
Catalog No. 50-287-9153 Supplier TARGETMOL CHEMICALS INC Supplier No. T829610MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 5,7,4'-Trimethoxyflavone (4',5,7-Trimethoxyflavone) is isolated from Kaempferia parviflora (KP) which is a medicinal plant from Thailand. Purity 97.53%