Learn More
TARGETMOL CHEMICALS INC 5 7 8-Trimethoxyflavone 10MG
SDP
Catalog No. 502875825
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg and bulk. Please contact Fisher for quotes. 5,7,8-Trimethoxyflavone (Norwogonin 5,7,8-trimethyl ether) is a compound extracted from *Andrographis paniculata*, exhibiting anti-inflammatory activity and NO inhibition. Purity 99.89%
Catalog No. 50-287-5825 Supplier TARGETMOL CHEMICALS INC Supplier No. TN199510MG
Also available in 1 mg, 5 mg, 25 mg and bulk. Please contact Fisher for quotes. 5,7,8-Trimethoxyflavone (Norwogonin 5,7,8-trimethyl ether) is a compound extracted from *Andrographis paniculata*, exhibiting anti-inflammatory activity and NO inhibition. Purity 99.89%