Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,7-bis(1,1-dimethylethyl)-3-hydroxy-3-(trifluoromethyl)-2(3H)-benzofuranone | 123557-91-5 | MFCD03764148 | 98.3% | 330.34 g/mol | C17H21F3O3 | 1 ML
SDP

Catalog No. 5000248873
Encompass_Preferred

rac-BHFF is a racemic positive allosteric modulator of the GABAB receptor used in research to potentiate GABAergic signaling. It is supplied in high-purity form for in vitro and in vivo pharmacology studies, available as a ready-to-use solution and as solid material for formulation or dosing flexibility.

  • Potent positive allosteric modulator of GABAB receptor.
  • Supplied as a 10 mM solution in DMSO and as solid quantities for flexibility.
  • High purity (≈98.3%), suitable for pharmacology research.
  • Molecular weight 330.34 g/mol and formula C17H21F3O3.
  • Useful for in vitro and in vivo studies of GABAergic signaling and receptor modulation.

Catalog No. 50-002-48873 Supplier Medchemexpress LLC Supplier No. HY10351910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

rac-BHFF is a racemic positive allosteric modulator of the GABAB receptor used in research to potentiate GABAergic signaling. It is supplied in high-purity form for in vitro and in vivo pharmacology studies, available as a ready-to-use solution and as solid material for formulation or dosing flexibility.

  • Potent positive allosteric modulator of GABAB receptor.
  • Supplied as a 10 mM solution in DMSO and as solid quantities for flexibility.
  • High purity (≈98.3%), suitable for pharmacology research.
  • Molecular weight 330.34 g/mol and formula C17H21F3O3.
  • Useful for in vitro and in vivo studies of GABAergic signaling and receptor modulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.